Order Entry
Netherlands
ContactUsLinkComponent
3-Nitro-o-xylene, Supelco®
3-Nitro-o-xylene, Supelco®
Catalog # 40870-25ML
Supplier:  Merck
CAS Number:  
undefined
3-Nitro-o-xylene, Supelco®
Catalog # 40870-25ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 mL
  • Seq:
    2315

Specifications

About this item

 :
 :151.16 g/mol
Boiling Pt:245 °C (1013 hPa)
Density:1.1402 g/cm³ (20 °C)
Storage Temperature:Ambient
ADR:6.1,II
 :M-Clarity™ quality level = MQ100
EINECS:201-474-3
UN:1665
CAS Number:83-41-0
Formula:C₈H₉NO₂
MDL Number:MFCD00007162