α,α,α,α',α',α'-Hexafluoro-m-xylene ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: 1,3-Bis(trifluoromethyl)benzene, α,α,α,β,β,β-Hexafluoro-m-xylene
CAS Number:402-31-3
:
Formula:C₈H₄F₆
MDL Number:MFCD00000392
Boiling Pt:115…117 °C
Melting Pt:–35 °C
Density:1.378
Molecular Weight:214.11 g/mol
Flash Pt:26 °C (78 °F)
Storage Temperature:Ambient
EINECS:206-939-4
UN:1993
ADR:3 III
:GHS02
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together