α-Chloro-p-xylene 98%
: alpha-Chloro-p-xylene, 4-Methylbenzyl chloride
:104-82-5
:
:C₈H₉Cl
:MFCD00000919
:200
:4 °C
:1.06 g/cm³
:140.61 g/mol
:75 °C
:3265
:8 II
:12,10224
:203-241-1
:GHS07
:GHS05
:Refrigerator
- Environmentally Preferable:
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5330 to 1.5370 (20°C, 589 nm)
- GC:≥97.5 %
Frequently Bought Together