Order Entry
Netherlands
ContactUsLinkComponent
Organic reference standard, Nitrosodiphenylamine 5,000 µg/ml in methanol, Supelco®
Organic reference standard, Nitrosodiphenylamine 5,000 µg/ml in methanol, Supelco®
Catalog # SUPL40060
Supplier:  Merck
CAS Number:  
Organic reference standard, Nitrosodiphenylamine 5,000 µg/ml in methanol, Supelco®
Catalog # SUPL40060
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1
  • CAS No.:
    86-30-6
  • Description:
    Nitrosodiphenylamine 5,000 µg/ml in methanol
  • Formula:
    C₁₂H₁₀N₂O
  • Description:
    Naphthalene

Specifications

About this item