Benzyldimethylhexadecylammonium chloride
Catalog # PROOMM0224.03
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
:396.10 g/mol
EINECS:204-526-3
CAS Number:122-18-9
Formula:C₂₅H₄₆ClN
MDL Number:MFCD00011625