0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Netherlands
ContactUsLinkComponent
Standards
Organic Standards
Acenaphthene-[D10] 2000 µg/ml in toluene, Dr Ehrenstorfer
Acenaphthene-[D10] 2000 µg/ml in toluene, Dr Ehrenstorfer
EHERYA20505100TO
:
EHRENSTORFER
:
Acenaphthene-[D10] 2000 µg/ml in toluene, Dr Ehrenstorfer
EHERYA20505100TO
:
EHRENSTORFER
:
Pack type:
Glass bottle
Pk:
1 mL
CAS No.:
15067-26-2
Description:
Acenaphthene-[D10] 2,000 µg/ml in toluene
Formula:
C₁₂D₁₀
Description:
Acenaphthene-[D10]