Order Entry
Netherlands
ContactUsLinkComponent
Organic reference standard, Triphenylene 10 µg/ml in acetonitrile, Dr Ehrenstorfer
Organic reference standard, Triphenylene 10 µg/ml in acetonitrile, Dr Ehrenstorfer
  EHERL20945000AL
 :  
Organic reference standard, Triphenylene 10 µg/ml in acetonitrile, Dr Ehrenstorfer
  EHERL20945000AL
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • CAS No.:
    217-59-4
  • Description:
    Triphenylene 10 µg/ml in acetonitrile
  • Formula:
    C₁₈H₁₂
  • Description:
    Triphenylene