Organic reference standard, 1-Methylnaphthalene 10 µg/ml in acetonitrile, Dr Ehrenstorfer
Catalog # EHERL20895000AL
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:90-12-0
- Description:1-Methylnaphthalene 10 µg/ml in acetonitrile
- Formula:C₁₁H₁₀
- Description:1-Methylnaphthalene