Pesticide reference standard, 2-Chloronaphthalene 10 µg/ml in isooctane, Dr Ehrenstorfer
EHERL20425200IO
:
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:91-58-7
- Description:2-Chloronaphthalene 10 µg/ml in isooctane
- Formula:C₁₀H₇Cl
- Description:2-Chloronaphthalene