Pesticide reference standard, 1-Chloronaphthalene 10 µg/ml in acetonitrile, Dr Ehrenstorfer
Catalog # EHERL20425100AL
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:90-13-1
- Description:1-Chloronaphthalene 10 µg/ml in acetonitrile
- Formula:C₁₀H₇Cl
- Description:1-Chloronaphthalene