Lidocaine
Catalog # EHERC14629790
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,1 g
- Seq:3020
Specifications
About this item
:
:234.34 g/mol
EINECS:205-302-8
CAS Number:137-58-6
Formula:C₁₄H₂₂N₂O
MDL Number:MFCD00026733