

Benzotrifluoride ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:98-08-8
:
Formula:C₇H₅F₃
MDL Number:MFCD00000372
:146.11 g/mol
Merck Index:14,1110
Boiling Pt:101…103 °C
Melting Pt:–29 °C
Density:1.188
Flash Pt:12 °C (54 °F)
Storage Temperature:Ambient
EINECS:202-635-0
UN:2338
ADR:3,II
:GHS02
:GHS09
Related Information
Frequently Bought Together