4-Chlorotoluene
Catalog # EHERC11522000
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
:126.59 g/mol
Merck Index:13,02190
Boiling Pt:162 °C (1013 hPa)
Melting Pt:6…8 °C
Density:1.07 g/cm³ (20 °C)
Flash Pt:51 °C
Storage Temperature:Ambient
EINECS:203-397-0
UN:2238
ADR:3,III
:GHS07
:GHS09
CAS Number:106-43-4
Formula:C₇H₇Cl
MDL Number:MFCD00000631