4-Chlorotoluene 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:106-43-4
:
Formula:C₇H₇Cl
MDL Number:MFCD00000631
:126.59 g/mol
Merck Index:13,02190
Boiling Pt:162 °C (760 mmHg)
Melting Pt:6…8 °C
Density:1.07 g/cm³
Flash Pt:51 °C
Storage Temperature:Ambient
EINECS:203-397-0
UN:2238
ADR:3,III
:GHS02
:GHS07
:GHS09
Specification Test Results
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Refractive index:1.5190 to 1.5210 (20°C, 589 nm)
- Water:≤0.01 %
- GC:≥97.5 %
- Color scale:≤15 APHA
Specification Test Results
Frequently Bought Together