
2-Bromo-4-(trifluoromethyl)aniline
Supplier: Thermo Scientific
Certificates
About this item
CAS Number:57946-63-1
:
Formula:C₇H₅BrF₃N
MDL Number:MFCD00042150
Boiling Pt:109…110 °C (10 mmHg)
Melting Pt:26…28 °C
Density:1.7 g/cm³ (20 °C)
EINECS:261-035-7
UN:2811
ADR:6.1,III
:240.02 g/mol
:GHS07
Frequently Bought Together