Tetraphenylethylene ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:632-51-9
:
Formula:C₂₆H₂₀
MDL Number:MFCD00004764
:332.44 g/mol
Boiling Pt:420 °C
Melting Pt:222…226 °C
Density:1.148 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:211-179-1
Related Information
Frequently Bought Together