Order Entry
Netherlands
ContactUsLinkComponent
4-Vinylbenzoic acid 96%
4-Vinylbenzoic acid 96%
Certificates
undefined
4-Vinylbenzoic acid 96%

Specification Test Results

  • Appearance:
    Cream to brown Powder
  • Infrared spectrum:
    Conforms
  • Solubility:
    (10% in Methanol) Clear solution
  • Titration with NaOH:
    ≥95 %

Specification Test Results

Related Information

Frequently Bought Together