(R)-(-)-2-Hydroxyisovaleric acid
Marketsource
Supplier: BACHEM BIOCHEMICA
Jump to:
About this item
Synonyms: D-α-Hydroxyisovaleric acid, 2-Hydroxyisovaleric-acid, (R)-2-Hydroxy-3-methylbutanoic acid, D-Valic acid, (2R)-2-Hydroxy-3-methylbutanoic acid
- CAS Number:17407-56-6
- Formula:C₅H₁₀O₃
- MDL Number:MFCD00066442
- Molecular Weight:118.13 g/mol
2 Options Available Below
Product Details & Documents
Synonyms: D-α-Hydroxyisovaleric acid, 2-Hydroxyisovaleric-acid, (R)-2-Hydroxy-3-methylbutanoic acid, D-Valic acid, (2R)-2-Hydroxy-3-methylbutanoic acid
- CAS Number:17407-56-6
- Formula:C₅H₁₀O₃
- MDL Number:MFCD00066442
- Molecular Weight:118.13 g/mol
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
- Specifications:MarketsourceMarketsource
- Specifications:MarketsourceMarketsource



