Order Entry
Netherlands
ContactUsLinkComponent
Organic reference standard, o-Anisidine 100 µg/ml in acetonitrile, Dr Ehrenstorfer
Organic reference standard, o-Anisidine 100 µg/ml in acetonitrile, Dr Ehrenstorfer
Catalog # EHERXA10266000AL
Supplier:  EHRENSTORFER
CAS Number:  
Organic reference standard, o-Anisidine 100 µg/ml in acetonitrile, Dr Ehrenstorfer
Catalog # EHERXA10266000AL
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    90-04-0
  • Description:
    o-Anisidine 100 µg/ml in acetonitrile
  • Formula:
    C₇H₉NO
  • Description:
    o-Anisidine

Specifications

About this item