

Palladium(II) sulphate dihydrate ≥99.95% (metals basis) Pd ≥ 44.1%, Premion®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13444-98-9
:
Formula:O₄PdS·2H₂O
MDL Number:MFCD00011173
Melting Pt: Dec.
Storage Temperature:Ambient
EINECS:236-957-8
UN:3260
ADR:8,III
:238.51 g/mol
:GHS05
:GHS07
Related Information
Frequently Bought Together