

5-Amino-1H-tetrazole ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:4418-61-5
:5378-49-4
Formula:CH₃N₅
MDL Number:MFCD00005248
:85.07 g/mol
Boiling Pt:387 °C (1013 hPa)
Melting Pt:216…217 °C
Density:1,51 g/cm³ (20 °C)
EINECS:224-581-7
UN:1325
ADR:4.1,III
:GHS02
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
Specification Test Results
Frequently Bought Together