Order Entry
Netherlands
ContactUsLinkComponent
α,α,α-Trifluoro-o-cresol ≥98.0% (by GC)
Corrosive
α,α,α-Trifluoro-o-cresol ≥98.0% (by GC)
Catalog # TCIAH0647-25G
Supplier:  TCI
CAS Number:  
undefined
Corrosive
α,α,α-Trifluoro-o-cresol ≥98.0% (by GC)
Catalog # TCIAH0647-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    44.0 to 47.0 deg-C

Specification Test Results

About this item

 :
Boiling Pt:149°C
Melting Pt:46 °C
Density:Min, 1 g/cm³ (20 °C)
ADR:8,III
 :162.11 g/mol
EINECS:207-148-7
 :GHS05
CAS Number:444-30-4
Formula:C₇H₅F₃O
MDL Number:MFCD00002222