2,6-Dichloropurine ≥97.0% (by HPLC, titration analysis)
: TCI
:5451-40-1
:
:C₅H₂Cl₂N₄
:MFCD00077725
:189 g/mol
:181 °C
:226-681-6
- Appearance:White to Light yellow to Dark green powder to crystal
- Purity (HPLC):min. 97.0 area%
- Purity (Neutralization titration):min. 97.0 %
Frequently Bought Together