Cleaning concentrate, liquid, rea-phos®
612-2622
- Description:rea-phos®, phosphate-free liqiuid concentrate
- Packed:Bottle
- Pk:1
Surface cleaner for all laboratory instruments made of glass, porcelain, plastic and metal (except aluminium and zinc). Particularly suitable for cleaning equipment used in the determination of phosphate levels such as water testing laboratories. Environmentally friendly, as the active agents.
- Phosphate-free rapid cleanser
- Similar in efficacy to laboratory cleaning agents containing phosphate
- Can be used with hard water
- Extremely economical: Typically 1 % working solution
- Suitable for use in ultrasonic cleaning baths