


2,4-Dinitroaniline
Catalog # EHERC12782000
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,1 g
- Seq:3020
Specifications
About this item
:
:183.12 g/mol
Merck Index:13,03298
Boiling Pt:401 °C (1013 hPa)
Melting Pt:177…178 °C
Density:1.6 g/cm³ (20 °C)
Flash Pt:224 °C
EINECS:202-553-5
UN:1596
ADR:6.1,II
:GHS06
:GHS08
:GHS09
Storage Temperature:Ambient
CAS Number:97-02-9
Formula:C₆H₅N₃O₄
MDL Number:MFCD00007151