


N,N-Dimethylaniline for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:121-69-7
:
Formula:C₈H₁₁N
MDL Number:MFCD00008304
:121.18 g/mol
Merck Index:13,03261
Boiling Pt:194 °C (1013 hPa)
Melting Pt:2.5 °C
Density:0.9559 g/cm³ (20 °C)
EINECS:204-493-5
UN:2253
ADR:6.1,II
:M-Clarity™ quality level = MQ200
Flash Pt:62 °C
:GHS06
:GHS08
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Solubility:1.2 g/l (20 °C)
- Melting Point:2.5 °C
- Molar Mass:121.18 g/mol
- Boiling Point:192 - 195 °C (1013 hPa)
- Vapor Pressure:0.53 hPa (20 °C)
- Flash Point:75 °C
- Refractive Index:1.5582 (20 °C, 589 nm)
- Explosion Limit:1.2 - 7 %(V)
- Density:0.96 g/cm3 (20 °C)
- pH:7.4 (1.2 g/l, H2O, 20 °C)
- Ignition Point:370 °C DIN 51794
- Synonyms:Dimethylaminobenzene
- Hill Formula:C8H11N
Specification Test Results
Frequently Bought Together