3,5-Diiodosalicylic acid
Catalog # MOLE28768172-100G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:389.92 g/mol
Merck Index:13,03212
Boiling Pt:381 °C (1013 hPa)
EINECS:205-124-0
:GHS07
Storage Temperature:Ambient
CAS Number:133-91-5
Formula:C₇H₄I₂O₃
MDL Number:MFCD00002444