3,5-Diiodosalicylic acid ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:133-91-5
:
Formula:C₇H₄I₂O₃
MDL Number:MFCD00002444
:389.92 g/mol
Merck Index:14,3190
Boiling Pt:381 °C (1013 hPa)
Melting Pt:228…234 °C
Storage Temperature:Ambient
EINECS:205-124-0
:GHS06
:GHS07
Related Information
Frequently Bought Together