
3-Iodobenzoic acid ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:618-51-9
:
Formula:C₇H₅IO₂
MDL Number:MFCD00002496
Boiling Pt:337 °C (1013 hPa)
Melting Pt:186…188 °C
Density:2.171 g/cm³ (20 °C)
:248.02 g/mol
Storage Temperature:Ambient
EINECS:210-555-2
:GHS07
Related Information
Frequently Bought Together