


1-Fluoro-2-nitrobenzene ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1493-27-2
:
Formula:C₆H₄FNO₂
MDL Number:MFCD00007048
:141.1 g/mol
Boiling Pt:215 °C
Melting Pt:–8 °C
Density:1.340
Flash Pt:94 °C (201 °F)
Storage Temperature:Ambient
EINECS:216-088-0
UN:2810
ADR:6.1,III
:GHS06
:GHS08
:GHS07
Related Information
Frequently Bought Together