

4-Fluoro-2-nitroaniline 99%
:364-78-3
:
:C₆H₅FN₂O₂
:MFCD00007830
:156.12 g/mol
:92…95 °C
:89 °C
:206-666-0
:GHS02
:GHS07
:Ambient
- Appearance:Orange to brown Crystalline powder
- Infrared spectrum:Conforms
- Melting point:92 to 95 °C
- GC:≥98.5 %
Frequently Bought Together