2,6-Difluoroaniline
Catalog # MOLE11813118-100G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:129.11 g/mol
Boiling Pt:51 °C (1013 hPa)
Density:1.199 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:226-853-0
UN:1992
ADR:3,III
:GHS02
:GHS07
CAS Number:5509-65-9
Formula:C₆H₅F₂N
MDL Number:MFCD00007655