2,6-Difluoroaniline 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:5509-65-9
:
Formula:C₆H₅F₂N
MDL Number:MFCD00007655
:129.11 g/mol
Boiling Pt:51…52 °C (15 mmHg)
Density:1.199 g/cm³
Flash Pt:43 °C
Storage Temperature:Refrigerator
EINECS:226-853-0
UN:1992
ADR:3,III
:GHS07
:GHS02
Specification Test Results
- Appearance:Clear yellow to brown Liquid (May darken on storage)
- Infrared spectrum:Conforms
- Refractive index:1.5070 to 1.5090 (20°C, 589 nm)
- GC:≥96.0 %
Specification Test Results
Frequently Bought Together