Pesticide reference standard, Pentachlorophenol 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalog # EHERL15970000CY
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:87-86-5
- Description:Pentachlorophenol 10 µg/ml in cyclohexane
- Formula:C₆Cl₅OH
- Description:Pentachlorophenol