


2,4,6-Trichlorophenol for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:88-06-2
:
Formula:C₆H₃Cl₃O
MDL Number:MFCD00002172
:197.45 g/mol
Merck Index:12,09773
Boiling Pt:244.5 °C (1013 hPa)
Melting Pt:∼66 °C
Density:1.675 g/cm³ (25 °C)
EINECS:201-795-9
UN:2020
ADR:6.1,III
:M-Clarity™ quality level = MQ200
Flash Pt:99 °C
:GHS08
:GHS07
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Solubility:0.43 g/l (20 °C)
- Melting Point:65 - 68 °C
- Molar Mass:197.44 g/mol
- Boiling Point:244 - 246 °C (1013 hPa)
- Vapor Pressure:0.035 hPa (20 °C)
- Flash Point:99 °C
- Density:1.675 g/cm3 (25 °C)
Specification Test Results
Frequently Bought Together