2,3,5-Trichlorophenol
Catalog # EHERC17773500
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,1 g
- Seq:3020
Specifications
About this item
:
:197.45 g/mol
Boiling Pt:248 °C (1013 hPa)
Melting Pt:62 °C
EINECS:213-272-2
UN:2020
ADR:6.1,III
CAS Number:933-78-8
Formula:C₆H₃Cl₃O
MDL Number:MFCD00002164