
2-Chlorobenzoic acid 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:118-91-2
:
Formula:C₇H₅ClO₂
MDL Number:MFCD00002412
:156.57 g/mol
Merck Index:13,02143
Boiling Pt:285 °C (760 mmHg)
Melting Pt:139…143 °C
Density:1,544 g/cm³ (20 °C)
Flash Pt:173 °C
EINECS:204-285-4
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to off-white Powder
- Infrared spectrum:Conforms
- Melting point:139 to 143 °C
- HPLC:≥97.5 %
- Titration with NaOH:≥97.5 %
- Copper (Cu):≤5 ppm
- Iron (Fe):≤10 ppm
- Lead (Pb):≤10 ppm
- Selenium (Se):≤10 ppm
- Zinc (Zn):≤10 ppm
- Heavy metals (as Pb):≤10 ppm
Specification Test Results
Frequently Bought Together