2,5-Dihydroxybenzoic acid for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:490-79-9
:
Formula:C₇H₆O₄
MDL Number:MFCD00002460
:154.12 g/mol
Merck Index:13,04409
Boiling Pt:339 °C (1013 hPa)
Melting Pt:202…206 °C
Density:1.55 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:207-718-5
Specification Test Results
- Melting Point:202 - 206 °C
- Molar Mass:154.12 g/mol
Specification Test Results
Frequently Bought Together