6-Nitro-m-anisic acid 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1882-69-5
:
Formula:C₈H₇NO₅
MDL Number:MFCD00151836
:197.15 g/mol
Melting Pt:125…130 °C
:GHS07
Specification Test Results
- Appearance:Beige to grey to brown Granules
- Infrared spectrum:Conforms
- Titration with NaOH:≥96.0 %
Specification Test Results
Frequently Bought Together