Order Entry
Netherlands
Orders LinkContactUsLinkComponent
4-(Dimethylamino)benzaldehyde, Supelco®

4-(Dimethylamino)benzaldehyde, Supelco®

Glass bottle, 500 mL
Catalog # 02560-500ML
Supplier:   Merck
4-(Dimethylamino)benzaldehyde, Supelco®
Catalog # 02560-500ML
Supplier:  Merck
CAS Number:  100-10-7
4-(Dimethylamino)benzaldehyde, Supelco®
4-(Dimethylamino)benzaldehyde, Supelco®

Glass bottle, 500 mL

CAS Number:  100-10-7
Price
304,00 €
Log in for availability
1 * 500 mL
Product Details & Documents
Synonyms:  Ehrlich's Reagent, 4-(N,N-Dimethylamino)benzaldehyde, p-Dimethylamino-Benzaldehyde, p-Dimethylaminobenzaldehyde, p-(Dimethylamino)benzaldehyde
  • CAS Number:100-10-7
  • Formula:(CH₃)₂NC₆H₄CHO
  • MDL Number:MFCD00003381
  • Molecular Weight:149.19 g/mol
  • Merck Index:13,03257
  • Boiling Pt:176…177 °C (1013 hPa)
  • Melting Pt:72 °C
  • Density:1.0254 g/cm³ (99.9 °C)
  •  :M-Clarity™ quality level = MQ100
  • Flash Pt:>100 °C
  • Storage Temperature:Ambient
  • EINECS:202-819-0
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.