1-Bromo-2-nitrobenzene 98%
:577-19-5
:
:C₆H₄BrNO₂
:MFCD00007045
:202.01 g/mol
:261 °C (760 mmHg)
:40…43 °C
:1,62 g/cm³ (20 °C)
:>110 °C
:Ambient
:209-409-0
:3459
:6.1,III
:GHS07
:GHS08
- Appearance:Yellow Fused solid or crystals and/or chunks
- Infrared spectrum:Conforms
- Melting point:39 to 43 °C
- GC:≥97.5 %
Frequently Bought Together