
2-Bromo-4-fluorobenzoic acid ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1006-41-3
:
Formula:C₇H₄BrFO₂
MDL Number:MFCD00055370
:219.01 g/mol
Boiling Pt:289 °C (1013 hPa)
Melting Pt:167…169 °C
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together