Pesticide reference standard, Pentachlorobenzene 100 µg/ml in cyclohexane, Dr Ehrenstorfer
EHERXA15960000CY
:
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:608-93-5
- Description:Pentachlorobenzene 100 µg/ml in cyclohexane
- Formula:C₆HCl₅
- Description:Pentachlorobenzene