Order Entry
Netherlands
ContactUsLinkComponent
Organic reference standard, m-Cresol 100 µg/ml in methanol, Dr Ehrenstorfer
Organic reference standard, m-Cresol 100 µg/ml in methanol, Dr Ehrenstorfer
Catalog # EHERXA15140300ME
Supplier:  EHRENSTORFER
CAS Number:  
Organic reference standard, m-Cresol 100 µg/ml in methanol, Dr Ehrenstorfer
Catalog # EHERXA15140300ME
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    108-39-4
  • Description:
    m-Cresol 100 µg/ml in methanol
  • Formula:
    C₇H₈O
  • Description:
    m-Cresol

Specifications

About this item