Organic reference standard, o-Cresol 100 µg/ml in methanol, Dr Ehrenstorfer
Catalog # EHERXA15140200ME
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:95-48-7
- Description:o-Cresol 100 µg/ml in methanol
- Formula:C₇H₈O
- Description:o-Cresol