Organic reference standard, 3-Methylfuran 100 µg/ml in methanol, Dr Ehrenstorfer
Catalog # EHERXA15086070ME
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:930-27-8
- Description:3-Methylfuran 100 µg/ml in methanol
- Formula:C₅H₆O
- Description:3-Methylfuran