Organic reference standard, Biphenyl (PCB No. 0) 100 µg/ml in methanol, Dr Ehrenstorfer
EHERXA10630000ME
:
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:92-52-4
- Description:Biphenyl (PCB No. 0) 100 µg/ml in methanol
- Environmentally Preferable:
- Formula:C₁₂H₁₀
- Description:Biphenyl (PCB No. 0)