Organic reference standard, 2-Methylphenanthrene 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalog # EHERL20900100CY
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:2531-84-2
- Description:2-Methylphenanthrene 10 µg/ml in cyclohexane
- Formula:C₁₅H₁₂
- Description:2-Methylphenanthrene