Organic reference standard, 2-Methylchrysene 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalog # EHERL20870000CY
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:3351-32-4
- Description:2-Methylchrysene 10 µg/ml in cyclohexane
- Formula:C₁₉H₁₄
- Description:2-Methylchrysene