Order Entry
Netherlands
ContactUsLinkComponent
Pesticide reference standard, Pentachlorobenzene 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Pesticide reference standard, Pentachlorobenzene 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalog # EHERL15960000CY
Supplier:  EHRENSTORFER
CAS Number:  
Pesticide reference standard, Pentachlorobenzene 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalog # EHERL15960000CY
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • CAS No.:
    608-93-5
  • Description:
    Pentachlorobenzene 10 µg/ml in cyclohexane
  • Formula:
    C₆HCl₅
  • Description:
    Pentachlorobenzene

Specifications

About this item